Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin alpha 3 Mouse anti-Human, Clone: 12C4, MilliporeSigma™

Catalog No. MABT413MI Shop All MilliporeSigma Products
missing translation for 'changeView'
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT413MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. MABT413MI Supplier MilliporeSigma missing translation for 'supplierNo' MABT413

Mouse Monoclonal Antibody

This Anti-Laminin alpha 3 antibody, clone 12C4 is validated for use in western blotting, immunocytochemistry and immunohistochemistry for the detection of Laminin alpha 3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin alpha 3
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 12C4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000218
Gene Symbols LAMA3, LAMNA
Host Species Mouse
Immunogen Recombinant protein corresponding to the C-terminus of human Laminin alpha 3.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.