Learn More
ECM Bioscience Laminin β2/γ1 Subunits Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience LM0461
Encompass_Preferred
Clone (M046) was generated from a recombinant human laminin 521 protein that includes a heterotrimer of three subunits laminin α5, laminin β2, and laminin γ1 subunits. Accession #P11047. Clone M046 detects native laminin isoforms that include both laminin β2 and γ1 subunits. In native immunoblots, the antibody detects laminin 521, laminin 221, and laminin 121, but does not detect lamin 332 and laminin 411. The antibody weakly detects laminin 511. Applications: ICC, WB(Nat.), E; Species Reactivity: Hu; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein G chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Laminin subunit gamma-1, Laminin B2 chain, S-laminin, S-LAM gamma, LAMC1, LAMB2, Laminin 521, Laminin beta 2
Catalog No. 50-206-8747
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.