Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP-1 (CD107a) Rat anti-Mouse, Clone: 1D4B, MilliporeSigma™

Catalog No. MABC39MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC39MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC39MI Supplier MilliporeSigma Supplier No. MABC39
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect LAMP-1 (CD107a) using this Anti-LAMP-1 (CD107a) Antibody, clone 1D4B validated for use in western blot, immunoprecipitation, immunocytochemistry, immunohistochemistry, flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAMP-1 (CD107a)
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1D4B
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_034814
Gene Symbols LAMP1, LAMP-1, LAMPA, LGP120, CD107a
Host Species Rat
Immunogen NIH/3T3 plasma membrane fraction of mouse LAMP-1.
Purification Method Protein G
Quantity 50 μL
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.