Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3-I/II Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. abc929mi Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC929MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC929MI Supplier MilliporeSigma Supplier No. ABC929
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-LC3-I/II Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of LC3-I/II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LC3-I/II
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit Polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols LC3; LC3A; MAP1A; MAP1BLC3A; MAP1LC3A; LC3I; LC3II
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LC3-I/II.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_115903
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.