Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lck (pY505) Mouse, Unlabeled, Clone: 4, BD

Catalog No. p-3878097 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612390 50 μg
BDB612391 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612390 Supplier BD Biosciences Supplier No. 612390
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Protein tyrosine phosphorylation is an essential step in the signal transduction cascade leading to T cell antigen receptor (TCR) activation. Lck is a protein kinase and a member of the src family of cytoplasmic protein-tyrosine kinases (PTKs). Members of this family have several common features: 1) unique N-terminal domains, 2) attachment to cellular membranes through a myristylated N-terminus, and 3) homologous SH2, SH3, and catalytic domains. The unique N-terminal domain of Lck interacts with the cytoplasmic tails of the CD4 and CD8 cell surface
glycoproteins. CD4 and CD8 bind to surface MHC class II and class I molecules, respectively. Lck is regulated by both kinases and phosphatases. Autophosphorylation at Y394 leads to conformational changes in the catalytic domain, which induces kinase activity. Repression of Lck occurs via phosphorylation at Y505, located near the carboxy-terminus. Phosphorylation of this tyrosine site is mediated by the Csk family of PTKs. Upon phosphorylation at this site, Lck associates with the SH2 domain in the amino-terminus, thus keeping the protein biologically inactive. Lck activity and regulation is critical for activation and development of T cells.

Flow Cytometry, Intracellular Staining, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Lck (pY505)
Applications Western Blot
Classification Monoclonal
Clone 4
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Lck (pY505)
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.