Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-LDL Receptor Clone: 15C8,

Catalog No. LP02100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
LP02100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LP02100UG Supplier MilliporeSigma™ Supplier No. LP02100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects LDL Receptor Clone: 15C8 in Bovine, Human samples, and it is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Immuno Blotting

The low density lipoprotein receptor is responsible for mediating the uptake of LDL, a cholesterol carrying plasma lipoprotein. The receptor is a member of a class of cell surface proteins which are concentrated in clathrin coated pits and carry external ligands into cells by receptor-mediated endocytosis. The purified receptor is a glycoprotein with a molecular weight of 160,000. It is synthesized as a precursor of molecular weight 120,000 that is then glycosylated to the mature form by addition of N-linked and O-linked sugars in the Golgi apparatus. Unlike other hepatic lectins, the orientation of the LDL-R is such that the amino terminus is to the exterior of the cell. It has recently been determined that the LDL-R has sequence homology with the epidermal growth factor receptor precursor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDL Receptor
Applications ELISA, Flow Cytometry, Immunoblot, Immunoblot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 15C8
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen a trypic digest of bovine low density lipoprotein receptor
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽160kDa LDL receptor protein in HT1080 and HepG2 cells.
Target Species Bovine, Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.