Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Lewis A (Blood Group Antigen) Ab-1, Clone: LWA01; same as 7LE, Epredia™ LabVision

Catalog No. p-3033468 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
100 μL
200 μL
5 Slides
500 μL
7 mL
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS494P0 100 μL 200μg/mL
MS494P1 500 μL 200μg/mL
MS494P 1 mL 200μg/mL
MS494P1ABX 100 μL 1mg/mL
MS494PABX 200 μL 1mg/mL
MS494R7 7 mL
MS494PCS 5 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS494P0 Supplier Epredia Supplier No. MS494P0

Mouse Monoclonal Antibody

Lewis A is a blood group antigen consisting of Gal Β1-3(Fuc α1-4) GlcNAc. Lewis A is found to be present primarily on epithelial cells such as colon and kidneys.2-4 In the tumors and dedifferentiated tissues, decrease of Lea antigen was observed.

Host Species: Mouse
Clone: LWA01; same as 7LE
Isotype: IgGa/κ
Cross Reactivity: Human. Does not react with rat. Others not-tested.
Epitope: Gal Β1-3(Fuc α1-4) GlcNAc
Positive Control: Colon
Cellular Localization: Cytoplasmic

Immunohistochemistry (Formalin/Paraffin)

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Antigen Lewis A (Blood Group Antigen) Ab-1
Classification Monoclonal
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Form Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.