Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Light Chain, lambda Mouse anti-Human, Alexa Fluor 647, Clone: JDC-12, BD

Catalog No. BDB563463 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563463 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563463 Supplier BD Biosciences Supplier No. 563463

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The JDC-12 monoclonal antibody specifically binds to human immunoglobulin light chain, lambda (μ). It does not not bind to immunoglobulin κ light chains or heavy chains.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Light Chain, lambda
Applications Flow Cytometry
Classification Monoclonal
Clone JDC-12
Conjugate Alexa Fluor 647
Description Ig λ; IGL@; IGL; Immunoglobulin lambda locus
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.