Learn More
Seracare Life Sciences Inc ANTI-LISTERIA SPECIES BIOTINA
Supplier:  Seracare Life Sciences Inc 53600030
Encompass
Affinity purified antibody isolated from a pool of serum from goats immunized with Listeria species was labeled with a biotin analog having a long carbon spacer arm. This minimizes steric hindrance and maximizes binding.
Catalog No. 50-672-93
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.