Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

lethal giant larvae homolog 1 (Drosophila), Mouse, Clone: 5G2, Abnova™

Catalog No. 89015833 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-015-833 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-015-833 Supplier Abnova Corporation Supplier No. H00003996M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant LLGL1.

This gene encodes a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq

Sequence: GIASCVFTRHGQGFYLISPSEFERFSLSARNITEPLCSLDINWPRDATQASYRIRESPKLSQANGTPSILLAPQSLDGSPDPAHSMGPDTPEPPEAALSP

Specifications

Antigen lethal giant larvae homolog 1 (Drosophila)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5G2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant LLGL1.
Formulation PBS with no preservative; pH 7.4
Gene LLGL1
Gene Accession No. NM_004140
Gene Alias DLG4/HUGL/HUGL-1/HUGL1/LLGL
Gene Symbols LLGL1
Host Species Mouse
Immunogen LLGL1 (NP_004131, 911 a.a. ∼ 1010 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3996
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.