Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRAT Mouse anti-Human, Mouse, Rat, Clone: M34-P1F10, MilliporeSigma™

Catalog No. MABN644MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN644MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN644MI Supplier MilliporeSigma Supplier No. MABN644

Mouse Monoclonal Antibody

Detect Lecithin retinol acyltransferase using this mouse monoclonal Anti-LRAT Antibody, clone M34-P1F10, validated for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western bBotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRAT
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone M34-P1F10
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1λ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_004735
Gene Symbols LRAT
Host Species Mouse
Immunogen Linear peptide corresponding to human LRAT.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.