Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP6 Ectodomain Mouse anti-Human, Clone: A59, MilliporeSigma™

Catalog No. MABS274MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS274MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS274MI Supplier MilliporeSigma Supplier No. MABS274
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-LRP6 antibody, clone A59, Ectodomain is validated for use in flow cytometry, immunoprecipitation and immunocytochemistry for the detection of LRP6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRP6
Applications Flow Cytometry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone A59
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002327
Gene Symbols LRP6
Host Species Mouse
Immunogen Purified protein corresponding to the ectodomain domain of human LRP6.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.