Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6G and Ly-6C Rat anti-Mouse, FITC, Clone: RB6-8C5, BD

Catalog No. p-3967631 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553126 0.1 mg
BDB553127 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB553126 Supplier BD Biosciences Supplier No. 553126
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The RB6-8C5 antibody reacts with a common epitope on Ly-6G and Ly-6C, previously known as the myeloid differentiation antigen Gr-1. In the bone marrow, the level of antigen expression is directly correlated with granulocyte differentiation and maturation.

Host Species: Rat
Clone: RB6-8C5
Isotype: IgG2b, κ
Species Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ly-6G and Ly-6C
Applications Flow Cytometry
Classification Monoclonal
Clone RB6-8C5
Concentration 0.5mg/mL
Conjugate FITC
Description Ly6c, Lymphocyte antigen 6C2; Lymphocyte antigen 6G, Ly6g, Gr-1
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Rat
Immunogen Not Reported
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.