Learn More
Bio X Cell InVivoMab anti-mouse CD4, Clone: YTS 191, Size: 100 MG
Supplier:  Bio X Cell BE0119R100MG
Encompass_Preferred
InVivoMab anti-mouse CD4, Clone: YTS 191, Size: 100 MG, Isotype: rat IgG2b, Applications: in vivo CD4+ T cell depletion
Catalog No. 50-139-2974
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.