Learn More
Bio X Cell InVivoPlus anti-mouse CD4, Clone: GK1.5, Size: 5 MG
Supplier:  Bio X Cell BP00031R005MG
Encompass_Preferred
InVivoPlus anti-mouse CD4, Clone: GK1.5, Size: 5 MG, Isotype: rat IgG2b , Applications: in vivo CD4+ T cell depletion, Flow cytometry
Catalog No. 50-139-3508
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.