Learn More
Bio X Cell InVivoMab anti-mouse CD8β (Lyt 3.2), Clone: 53-5.8, Size: 5 MG
Supplier:  Bio X Cell BE0223R005MG
Encompass
InVivoMab anti-mouse CD8β (Lyt 3.2), Clone: 53-5.8, Size: 5 MG, Isotype: rat IgG1, Applications: in vivo CD8+ T cell depletion, in vitro CD8 blockade, Immunofluorescence
Catalog No. 50-139-3249
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.