Learn More
Bio X Cell InVivoMab anti-mouse IL-17A, Clone: 17F3, Size: 5 MG
Supplier:  Bio X Cell BE0173R005MG
Encompass_Preferred
InVivoMab anti-mouse IL-17A, Clone: 17F3, Size: 5 MG, Isotype: mouse IgG1, Applications: in vivo IL-17A neutralization
Catalog No. 50-562-545
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.