Learn More
Bio X Cell InVivoMab anti-mouse IL-6R, Clone: 15A7, Size: 5 MG
Supplier:  Bio X Cell BE0047R005MG
Encompass_Preferred
InVivoMab anti-mouse IL-6R, Clone: 15A7, Size: 5 MG, Isotype: rat IgG2b, Applications: in vivo blocking of IL-6/IL-6R signaling, in vitro blocking of IL-6R signaling
Catalog No. 50-562-144
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.