Learn More
Bio X Cell InVivoPlus anti-mouse PD-L1 (B7-H1), Clone: 10F.9G2, Size: 5 MG
Supplier:  Bio X Cell BP0101R005MG
Encompass
InVivoPlus anti-mouse PD-L1 (B7-H1), Clone: 10F.9G2, Size: 5 MG, Isotype: rat IgG2b, Applications: in vivo PD-L1 blockade, Immunofluorescence, Immunohistochemistry (frozen), Flow cytometry
Catalog No. 50-139-3590
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.