Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mac-3 Rat anti-Mouse, Unlabeled, Clone: M3/84, BD

Catalog No. bdb550292 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550292 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550292 Supplier BD Biosciences Supplier No. 550292
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The M3/84 antibody reacts with the 110kDa Mac-3 antigen expressed on mouse mononuclear phagocytes. Surface expression of the 92 to 110kDa glycoprotein antigen increases during differentiation of monocytes to activated peritoneal macrophages. The M3/84 mAb can detect CD107b antigen on tissue macrophages, thioglycollate-elicited peritoneal macrophages, and some myeloid cell lines, but not on lymphocytes or monocytes. In the bone marrow, very few cells display CD107b antigen on the surface, but a large proportion express cytoplasmic CD107b. The M3/84 antibody has also been reported to stain dendritic cells and endothelium in sections of thymus (both medulla and cortex), lymph nodes, spleen (white pulp), and gut-associated lymphoid tissue.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mac-3
Applications Flow Cytometry
Classification Monoclonal
Clone M3/84
Concentration 31.25μg/mL
Conjugate Unconjugated
Description CD107b; Lamp2; LAMP-2; Lysosome-associated membrane glycoprotein 2; LGP-B
Formulation Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse C57Bl/6 peritoneal exudate cells
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.