Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAEL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520923
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520923 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520923 Supplier Thermo Scientific Supplier No. PA520923
Only null left
Add to Cart
Add to Cart

MAEL Polyclonal Antibody

MAEL Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen MAEL
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MAEL
Gene Accession No. Q8BVN9, Q96JY0
Gene Alias 4933405K18Rik; AU019877; cancer/testis antigen 128; CT128; MAEL; maelstrom homolog; maelstrom homolog (Drosophila); maelstrom spermatogenic transposon silencer; protein maelstrom homolog; RGD1309333; RP11-102C16.1; SPATA35; spermatogenesis associated 35; testicular tissue protein Li 116
Gene Symbols MAEL
Host Species Rabbit
Immunogen A 16 amino acid peptide near the carboxy terminus of human Maelstrom.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 364039, 84944, 98558
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.