Learn More
Maltose Binding Protein Rabbit anti-Bacteria, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Maltose Binding Protein |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Rabbit serum. Liquid. Contains 0.02% sodium azide. |
| Host Species | Rabbit |
| Immunogen | MBP, purified recombinant. |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.