Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MAP3K3, MAP2K5 Protein Protein Interaction Antibody Pair, Abnova™

Catalog No. 89023308 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-023-308 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-023-308 Supplier Abnova Corporation Supplier No. DI0549
Add to Cart
Add to Cart

Includes

MAP3K3 rabbit purified polyclonal antibody (20μg); MAP2K5 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the MAP3K3 protein, and the other against the MAP2K5 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Antigen MAP3K3
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene MAP3K3
Gene Alias MAPKKK3/MEKK3
Gene Symbols MAP3K3
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4215/5607
Includes Antibody pair set content: 1. MAP3K3 rabbit purified polyclonal antibody (20 ug) 2. MAP2K5 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type MAP3K3 & MAP2K5 Protein Protein Interaction Antibody Pair
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.