Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MAPK14, MAPKAPK3 Protein Protein Interaction Antibody Pair, Abnova™

Catalog No. 89023103 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-023-103 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-023-103 Supplier Abnova Corporation Supplier No. DI0344
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

MAPK14 rabbit purified polyclonal antibody (20μg); MAPKAPK3 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the MAPK14 protein, and the other against the MAPKAPK3 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Antigen MAPK14
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene MAPK14
Gene Alias CSBP1/CSBP2/CSPB1/EXIP/Mxi2/PRKM14/PRKM15/RK/SAPK2A/p38/p38ALPHA
Gene Symbols MAPK14
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1432/7867
Includes Antibody pair set content: 1. MAPK14 rabbit purified polyclonal antibody (20 ug) 2. MAPKAPK3 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type MAPK14 & MAPKAPK3 Protein Protein Interaction Antibody Pair
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.