Learn More
Invivogen Anti-mDectin-1-IgG Monoclonal Antibody, Targets Murine Dectin-1, Purified by Affinity Chromatography, 2 x 100 µg
SDP
Supplier:  Invivogen MABGMDECT2
Encompass
Anti-mDectin-1-IgG (mDectin-1) is a monoclonal rat IgG2a antibody designed for detecting and neutralizing murine Dectin-1, crucial for antifungal innate immunity. It offers high specificity with no cross-reactivity to human Dectin-1 and is validated for use in neutralization assays and flow cytometry. This product is lyophilized and azide-free, ensuring optimal performance and safety in research applications. Storage is at -20°C, maintaining stability and efficacy.
Catalog No. NC2684392
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.