Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mena Mouse anti-Rat, Human, Mouse, Clone: A351F7D9, MilliporeSigma™

Catalog No. MAB2635MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2635MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2635MI Supplier MilliporeSigma Supplier No. MAB2635
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Mena using this Anti-Mena Antibody, clone A351F7D9 validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mena
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone A351F7D9
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001008493
Gene Symbols MENAH, ENAH, NDPP1, ENA
Host Species Mouse
Immunogen Histidine-tagged recombinant protein corresponding to human Mena and an additional fragment of Mena +, comprising only the + exon
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.