Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Metapneumovirus Mouse anti-Human, Clone: 134, MilliporeSigma™

Catalog No. MAB80125MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB80125MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB80125MI Supplier MilliporeSigma Supplier No. MAB80125
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Human Metapneumovirus Antibody, clone 134 is validated for use in ELISA, flow cytometry, immunofluorescence for the detection of Human Metapneumovirus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Metapneumovirus
Applications ELISA, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 134
Conjugate Unconjugated
Formulation Liquid in 0.02M PB with 0.25M NaCl, pH 7.6. Contains 0.1% sodium azide.
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.