Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR1 Mouse, Unlabeled, Clone: 20, BD

Catalog No. p-3851784 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610964 50 μg
BDB610965 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610964 Supplier BD Biosciences Supplier No. 610964
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glutamate is a major excitatory neurotransmitter and functions in multiple roles in the CNS. The functional diversity of glutamate is exemplified by two distinct groups of glutamate receptors: ionotropic and metabotropic. Coupling with G proteins provides the metabotropic glutamate receptors (mGluRs) with the capacity for intracellular signal transduction. Eight metabotropic glutamate receptors (mGluR1-8) and several Ca2+ sensing receptors belong to a novel G-protein coupled receptor (GPCR) family. The mGluRs possess the seven putative transmembrane domains which are characteristic of GPCR proteins. However, they exhibit no additional sequence homology to any member of other GPCR families. mGluR1 has large hydrophilic sequences in both the N- and C-terminal sides of the seven transmembrane domains. The sizable extracellular N-terminal domain is homologous to bacterial periplasmic binding proteins and serves as the glutamate binding site. mGluR1 activates phospholipase C (PLC), resulting in phosphoinositide turnover and, in turn, Ca2+; mobilization necessary for many signal transduction events.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen mGluR1
Applications Western Blot
Classification Monoclonal
Clone 20
Concentration 250μg/mL
Conjugate Unconjugated
Description Metabotropic Glutamate Receptor-1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat mGluR1 aa. 1042-1160
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.