Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-MHC class II (I-A/I-E), APC, Clone: M5/114.15.2,

Catalog No. MABF805MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF805MI 25 μg
MABF806MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF805MI Supplier MilliporeSigma™ Supplier No. MABF805
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects MHC class II (I-A/I-E) Clone: M5/114.15.2 in Mouse samples, and it is validated for Flow Cytometry

The M5/114.15.2 antibody reacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, as well as being cross-reactive with mouse cells of H-2p and H-2r haplotype. MHC Class II is widely expressed by mouse immune cells bearing these alloantigens, including T and B cells, monocytes, macrophages, and dendritic cells. The antibody does not react with the following alloantigens: I-Af, I-Ak, I-As, or NOD H-2g7.The M5/114.15.2 antibody may be used for analysis of mouse cells expressing MHC Class II alloantigens as described. Please note that the M5/114.15.2 clone may also be referred to as M5/114 in the literature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC class II (I-A/I-E)
Applications Flow Cytometry
Classification Monoclonal
Clone M5/114.15.2
Conjugate APC
Formulation Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Gene Accession No. P04441
Gene Symbols CD74, Ii
Host Species Rat
Immunogen Proprietary immunogen
Purification Method Purified by size-exclusion chromatography
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation, Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.