Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mint3 Mouse, Unlabeled, Clone: 32, BD

Catalog No. BDB611380 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611380 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611380 Supplier BD Biosciences Supplier No. 611380
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Munc18-1-interacting proteins, Mint1, Mint2, and Mint3, are members of the X11 family of proteins, which contain a phosphotyrosine-binding (PTB) domain and two PSD-95-DLG-ZO-1 (PDZ) domains. The PTB domain binds Asn-Pro-X-pTyr, a β-turn motif found on activated growth factor receptors and other signaling molecules. PDZ domains bind the C-terminus of proteins involved in receptor and channel clustering and protein localization in polarized cells. Both Mint1 and Mint2 are expressed primarily in the nervous system and may interact with Munc18-1 and syntaxin to form a multimeric complex that mediates appropriate docking/fusion of synaptic vesicles. In addition, both Mint1 and Mint2 may be involved in Alzheimer's disease, since Mint2 colocalizes with amyloid precursor protein (APP) and is found in neuritic plaques, while Mint1 can bind APP, and inhibits the processing of APP to the amyloid β peptide. Mint3 can also bind APP, but differs from Mint1 and Mint2 in its N-terminal region. It is also more widely expressed. Thus, Mint3 may function in signaling pathways, vesicle exocytosis, and/or protein targeting in a wide range of tissues. Mint3 has a calculated molecular weight of 61kDa , but reportedly is observed migrating at 86-89kDa.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mint3
Applications Western Blot
Classification Monoclonal
Clone 32
Concentration 250μg/mL
Conjugate Unconjugated
Description X11γ
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Mint3 aa. 63-185
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.