Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLKL Rat anti-Horse, Human, Mouse, Clone: 3H1, MilliporeSigma™

Catalog No. MABC604MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC604MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC604MI Supplier MilliporeSigma Supplier No. MABC604
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Anti-MLKL Antibody, clone 3H1 is validated for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting for the detection of MLKL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MLKL
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3H1
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG1 in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_083281
Gene Symbols Mlkl
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to mouse MLKL.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.