Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monkey IgM, Isotype Control, Novus Biologicals™

Monkey Antibody
Supplier: Novus Biologicals NBP197060
Description
IgM specifically detects IgM in Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, SDS-Page.Specifications
| IgM | |
| Unconjugated | |
| 0.1 M Tris Chloride, 0.5 M Sodium Chloride, pH 8.0. Stabilizer: None. with 0.1% Sodium Azide | |
| Monkey | |
| 0.25 mg | |
| Isotype Control (Primary) | |
| No reaction was observed against anti-Monkey IgG F(c). Some light chain cross reactivity will occur with anti-Monkey IgG. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgM |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), SDS-Page | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, SDS-Page 1:100 - 1:2000 | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Multi-step | |
| RUO | |
| 3507 | |
| Monkey | |
| Concentrated |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction