Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monocyte/Granulocyte Mouse anti-Pig, Unlabeled, Clone: 74-22-15A, BD

Catalog No. BDB553640 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553640 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553640 Supplier BD Biosciences Supplier No. 553640
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 74-22-15A (switch variant of 74-22-15) antibody, an isotype class-switch variant of mAb 74-22-15, reacts with a 230kDa protein1 expressed by most macrophages, peripheral blood monocytes and granulocytes, and few lymphocytes. mAb 74-22-15A does not cross-react with human or bovine cells. This clone was clustered as anti-SWC3a at the First International Swine CD workshop.

Host Species: Mouse
Clone: 74-22-15A
Isotype: IgG2b, κ
Species Reactivity: Pig
Immunogen: dd miniature swine thymocytes

Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen Monocyte/Granulocyte
Applications Flow Cytometry
Classification Monoclonal
Clone 74-22-15A
Concentration 0.5mg/mL
Conjugate Unconjugated
Description SWC3; CD172a
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen dd miniature swine thymocytes
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Pig
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.