Learn More
STEMCELL Technologies Anti-Mouse CD11c Antibody, Clone N418, APC, Size: 25 µg
SDP
Supplier:  STEMCELL Technologies 60002AZ.1
Encompass_Preferred
Clone N418 is a hamster (Armenian) monoclonal antibody that reacts with mouse CD11c (αX integrin), a 150 kDa type 1 transmembrane glycoprotein that associates non-covalently with CD18 (β2 integrin) to form a heterodimeric cell surface adhesion receptor. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of CD11c. Through its interaction with ligands such as iC3b, fibrinogen, and CD54, the CD11c/CD18 receptor is involved in several immune response processes, including cell migration, stimulation of cytokine production by monocytes and macrophages, T cell proliferation, leukocyte recruitment, and phagocytosis. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD11c Positive Selection Kit II (Catalog #18780). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-4880
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.