Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse CD140A, Brilliant Violet™ 421, Clone: APA5; BD

Catalog No. BDB562774 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562774 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562774 Supplier BD Biosciences Supplier No. 562774
Add to Cart
Add to Cart

Host Species: Rat
Clone: APA5
Isotype: IgG2a, κ
Cross Reactivity Species: Ms
Immunogen: Mouse PDGF Receptor α chain

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse CD140A
Classification Monoclonal
Clone APA5
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Host Species Rat
Immunogen Mouse PDGF Receptor α chain
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.