Learn More
STEMCELL Technologies Anti-Mouse CD19 Antibody, Clone 6D5, PE, Size: 50 µg
SDP
Supplier:  STEMCELL Technologies 60006PE.1
Encompass
Clone 6D5 is a rat monoclonal antibody that reacts with mouse CD19, an ~95 kDa type 1 transmembrane glycoprotein expressed on the surface of B cells throughout all stages of development, from early pre-B cells to plasma cells. Verified for use with cell separation and flow cytometry assays, use this antibody in applications requiring the detection of CD19. Activation of CD19 is accompanied by phosphorylation of the cytoplasmic domain, which promotes binding to kinases and the induction of intracellular signaling cascades. Mutations in CD19 can result in severe immunodeficiency syndromes. Clone 6D5 recognizes the same epitope as clone ID3 in cross-competition assays. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse B Cell Isolation Kit (Catalog #19854). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-4931
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.