Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
This product is suitable for:
- Sensitizing cells in hemolytic complement assays.
- Lymphocyte screening procedures, especially screening for T cells and T receptor-bearing lymphocytes.
- Use as a reagent in enzyme immunoassays (EIA)
- Cell and tissue staining (for light microscopy).
- Cell and tissue labeling (for electron microscopy).
- Blot immunostaining.
Note: F(ab')2 fragments are recommended for staining of cells or tissues which contain Fc receptors.
Preparation Method
- Antibody and highly purified HRP (Rz>3.0) are conjugated under defined conditions to obtain optimally labeled product.
- Conjugated protein is purified by salt fractionation.
- The product is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, adjusted to standard titer, filtered through a 0.22 μm filter, vialed and lyophilized.
- Titer: 1:78,000
- No preservative has been added.
Specifications
Specifications
| Applications | Immunoassay, Immunoblot |
| Classification | Polyclonal |
| Concentration | 19.0 mg/mL |
| Conjugate | Peroxidase |
| Formulation | Lyophilized |
| Host Species | Goat |
| Purification Method | Purified |
| Quantity | 2 mL |
| Test Specificity | It shows reactivity to mouse complement C3; cross-reactivity to other species may exist. Antibody activity to other serum proteins is not present. |
| Target Species | Mouse |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?