Learn More
Selleck Chemical LLC Anti-mouse IL-6R-InVivo
SDP
Supplier:  Selleck Chemical LLC A21575MG
Encompass_Preferred
Anti-mouse IL-6R-InVivo reacts with the mouse IL-6 receptor chain also known as CD126 CD126 is an 80 kDa type I cytokine receptor and a member of the immunoglobulin superfamily
Catalog No. 50-313-5868
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.