Learn More
ProteinSimple Anti-Mouse Detection Module, 1ea
Supplier:  ProteinSimple DM002
Encompass
Use with a Wes or Peggy Sue/Sally Sue a Separation Module. Includes: Luminol-S, Peroxide, Antibody Diluent 2, Anti-Mouse Secondary Antibody, Streptavidin-HRP.
Catalog No. NC1178181