Learn More
STEMCELL Technologies Anti-Mouse F4/80 Antibody, Clone BM8, APC, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60027AZ
Encompass
Clone BM8 is a rat monoclonal antibody that reacts with the F4/80 antigen (also termed Ly-71 in mouse and EMR-1 in human), an ~160 kDa transmembrane glycoprotein belonging to the EGF-TM7 family of G-protein-coupled receptors. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of human and mouse F4/80. F4/80 is reportedly the only macrophage marker suitable for distinguishing destructive from non-destructive inflammatory processes in the pancreas. The protein is thought to play a role in the generation of CD8+ regulatory T cells involved in immune tolerance. This antibody clone has also been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Monocyte Enrichment Kit (Catalog #19761) and EasySep™ Mouse CD11b Positive Selection Kit (Catalog #18770). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5131
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.