Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse VISTA/B7-H5/PD-1H Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. FAB7005G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7005G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7005G Supplier R&D Systems Supplier No. FAB7005G
This item is not returnable. View return policy

Sheep Polyclonal Antibody

VISTA/B7-H5/PD-1H Polyclonal specifically detects VISTA/B7-H5/PD-1H in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen VISTA/B7-H5/PD-1H
Applications Flow Cytometry
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Gene Accession No. Q9D659
Gene Alias 4632428N05Rik, B7H5, B7-H5, C10orf54, chromosome 10 open reading frame 54, Dies1, Gi24, PD1H, PD-1H, platelet receptor Gi24, PP2135, SISP1, stress induced secreted protein 1, VSIR
Gene Symbols Vsir
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Gi24/VISTA/B7-H5 Phe33-Ala191 Accession # Q9D659
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64115
Test Specificity Detects mouse Gi24/VISTA/B7-H5 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human Gi24/VISTA/B7-H5 is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.