Learn More
Rockland Immunochemicals Anti-MOUSE IgG (H&L) (GOAT) Antibody DyLight™ 800 Conjugated (5 X 100 µg) - 610-145-002-0.5
SDP
Supplier:  Rockland Immunochemicals 6101450020.5
Encompass_Preferred
Anti-Mouse IgG DyLight 800 Antibody generated in goat detects reactivity to Mouse IgG. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Rabbit, Rat and Sheep Serum Proteins.  This antibody will react with heavy chains of mouse IgG and with light chains of most mouse immunoglobulins. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms. The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Catalog No. 50-105-8408
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.