Learn More
Medchemexpress LLC Anti-Mouse Ter-119 A 10mg | 10mg
Supplier: Medchemexpress LLC HYP99015810MG
Anti-Mouse Ter-119 Antibody (TER-119) is an anti-mouse Ter-119 IgG2b monoclonal antibody Anti-Mouse Ter-119 Antibody (TER-119) can increase platelet count Anti-Mouse Ter-119 Antibody (TER-119) can remove red blood cells and their precursor cells to isolate and enrich natural killer (NK) cells Anti-Mouse Ter-119 Antibody (TER-119) can be used for research on immune thrombocytopenia Anti-Mouse Ter-119 Antibody (TER-119) can be used to construct a model of immune thrombocytopenic purpura combined with CD41 mAb[1][2][3]
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.