Learn More
Medchemexpress LLC Anti-Mouse Ter-119 A 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9901581MG
Encompass_Preferred
Anti-Mouse Ter-119 Antibody (TER-119) is an anti-mouse Ter-119 IgG2b monoclonal antibody Anti-Mouse Ter-119 Antibody (TER-119) can increase platelet count Anti-Mouse Ter-119 Antibody (TER-119) can remove red blood cells and their precursor cells to isolate and enrich natural killer (NK) cells Anti-Mouse Ter-119 Antibody (TER-119) can be used for research on immune thrombocytopenia Anti-Mouse Ter-119 Antibody (TER-119) can be used to construct a model of immune thrombocytopenic purpura combined with CD41 mAb[1][2][3]
Catalog No. 50-003-25283