Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

membrane protein, palmitoylated 4 (MAGUK p55 subfamily member 4), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004699 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-699 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-699 Supplier Abnova Corporation Supplier No. H00058538A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant MPP4.

This gene encodes a member of the membrane-associated guanylate kinase (MAGUK) protein family, with an N-terminal PDZ domain, a central src homology 3 region (SH3), and a C-terminal guanylate kinase-like (GUK) domain. The protein is localized to the outer limiting membrane in the retina, and is thought to function in photoreceptor polarity and the organization of specialized intercellular junctions. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq

Sequence: MIQSDKGADPPDKKDMKLSTATNPQNGLSQILRLVLQELSLFYSRDVNGVCLLYDLLHSPWLQALLKIYDCLQEFKEKKLVPATPHAQVLSYEVVELLRETPTSPEIQE

Specifications

Antigen membrane protein, palmitoylated 4 (MAGUK p55 subfamily member 4)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant MPP4.
Formulation 50% glycerol
Gene MPP4
Gene Accession No. NM_033066
Gene Alias ALS2CR5/DLG6
Gene Symbols MPP4
Host Species Mouse
Immunogen MPP4 (NP_149055, 1 a.a. ∼ 109 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 58538
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.