Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mRNA turnover 4 homolog (S. cerevisiae), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018474 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-474 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-474 Supplier Abnova Corporation Supplier No. H00051154D01P
Only null left
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human MRTO4 protein.

This gene encodes a protein sharing a low level of sequence similarity with ribosomal protein P0. While the precise function of the encoded protein is currently unknown, it appears to be involved in mRNA turnover and ribosome assembly. [provided by RefSeq

Sequence: MPKSKRDKKVSLTKTAKKGLELKQNLIEELRKCVDTYKYLFIFSVANMRNSKLKDIRNAWKHSRMFFGKNKVMMVALGRSPSDEYKDNLHQVSKRLRGEVGLLFTNRTKEEVNEWFTKYTEMDYARAGNKAAFTVSLDPGPLEQFPHSMEPQLRQLGLPTALKRGVVTLLSDYEVCKEGDVLTPEQARVLKLFGYEMAEFKVTIKYMWDSQSGRFQQMGDDLPESASESTEESDSEDDD

Specifications

Antigen mRNA turnover 4 homolog (S. cerevisiae)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human MRTO4 protein.
Formulation PBS with no preservative; pH 7.4
Gene MRTO4
Gene Accession No. NM_016183
Gene Alias C1orf33/MRT4/dJ657E11.4
Gene Symbols MRTO4
Host Species Rabbit
Immunogen MRTO4 (NP_057267.2, 1 a.a. ∼ 239 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51154
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.