Learn More
Leinco Technologies Inc Anti-Mouse CD90 (Thy-1) In Vivo PLATINUM; Clone HK2.1; Isotype: Rat IgG2c
SDP
Supplier:  Leinco Technologies Inc C6351.0 MG
Encompass_Preferred
Specificity: Clone HK2.1 recognizes Thy-1.1 & Thy-1.2 alloantigens of all mouse strains tested. Antigen Distribution: CD90 is expressed on hematopoietic stem cells, thymocytes, a subset of peripheral blood T cells, early myeloid & erythroid progenitors, immature B cells, neurons, activated endothelium, mast cells, dendritic cells & glomeruli. Background: CD90 is a 28-30 kD GPI-linked membrane glycoprotein & is part of the Ig superfamily. It interacts with CD45 in signal transduction during lymphocyte proliferation & differentiation. CD90 mediates adhesion of thymocytes to thymic stroma. Applications: (WB, FA) Clone HK2.1 has been implicated in activation of T-cells. Leinco’s In vivo GOLDTM & In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques & are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6041
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.