Learn More
Seracare Life Sciences Inc ANTI-MS IGG (H+L) AB XSA HRP
Supplier:  Seracare Life Sciences Inc 52200338
Encompass_Preferred
Affinity purified polyclonal antibody to mouse IgG, heavy and light chain specific (whole IgG), made in goat and labeled wih horseradish peroxidase. Product has been cross-adsorbed to bovine, chicken., goat, guinea pig, hamster, horse, human, rabbit, rat and sheep to minimize cross reactivity to these species. Product is in lyophhilized form.
Catalog No. 50-672-06
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.