Learn More
Leinco Technologies Inc Anti-Mouse LPAM-1 (LPAM1, Integrin α4β7, CD49d/β7, ITGA4, ITGB7) In Vivo PLATINUM; Clone DATK32; Isotype: Rat IgG2a κ
SDP
Supplier:  Leinco Technologies Inc L6075.0 MG
Encompass_Preferred
Specificity: Clone DATK32 is specific for LPAM1. Antigen distribution: LPAM1 is expressed on the majority of peripheral lymphocytes, subsets of thymocytes and bone marrow cells. Background: LPAM1 is a heterodimer comprised of a 130 kDa integrin β7 chain and a 150 kDa integrin α4 chain, and is a member of the Ig superfamily. Applications: (WB, B) Clone DATK32 induces LPAM1-dependent lymphocyte aggregation. DATK32 has been reported to block LPAM1-mediated cell adhesion in vivo, including binding to fibronectin, MAdCAM-1, and VCAM-1. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6531
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.