Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

microseminoprotein, beta-, Mouse, Clone: 3B11, Abnova™

Catalog No. 89000814 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-814 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-814 Supplier Abnova Corporation Supplier No. H00004477M08
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MSMB.

The protein encoded by this gene is a member of the immunoglobulin binding factor family. It is synthesized by the epithelial cells of the prostate gland and secreted into the seminal plasma. This protein has inhibin-like activity. It may have a role as an autocrine paracrine factor in uterine, breast and other female reproductive tissues. The expression of the encoded protein is found to be decreased in prostate cancer. Two alternatively spliced transcript variants encoding different isoforms are described for this gene. The use of alternate polyadenylation sites has been found for this gene. [provided by RefSeq

Sequence: MNVLLGSVVIFATFVTLCNASCYFIPNEGVPGDSTRKCMDLKGNKHPINSEWQTDNCETCTCYETEISCCTLVSTPVGYDKDNCQRIFKKEDCKYIVVEKKDPKKTCSVSEWII

Specifications

Antigen microseminoprotein, beta-
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3B11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant MSMB.
Formulation PBS with no preservative; pH 7.4
Gene MSMB
Gene Accession No. BC005257
Gene Alias HPC13/IGBF/MSP/MSPB/PN44/PRPS/PSP/PSP-94/PSP57/PSP94
Gene Symbols MSMB
Host Species Mouse
Immunogen MSMB (AAH05257.1, 1 a.a. ∼ 114 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4477
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.